Round-Bottom Polypropylene Test Tubes With Cap
Round-Bottom Polypropylene Test Tubes With Cap
Actual product may vary
Round-Bottom Polypropylene Test Tubes With Cap
Round-Bottom Polypropylene Test Tubes With Cap
Round-Bottom Polypropylene Test Tubes With Cap
Round-Bottom Polypropylene Test Tubes With Cap
Round-Bottom Polypropylene Test Tubes With Cap
Fisherbrand™'s round bottom tubes provide reliable containment for various laboratory fluid samples and preparations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)SterilityGraduatedPackaging
FISFB149569C14 mLSterileYesBlister Pack
FISFB149561D5 mLSterileNoBlister Pk.
Catalog number FISFB149569C
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
14 mL
Sterility:
Sterile
Graduated:
Yes
Packaging:
Blister Pack
  • Ideal for use in laboratory protocols requiring round bottom tubes
  • Ensures reliable handling of liquid samples
  • Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability, while polystyrene tubes offer high optical clarity
  • Available with and without a snap cap - dual-position snap caps offer both vented and fully closed positions.
  • 3,000 RCF rating for polypropylene tubes, 1,400 RCF rating for polystyrene tubes
  • Dimensions and volumes are approximate
Specifications
ColorTranslucent
Capacity (Metric)14 mL
Length (Metric)100 mm
MaterialPolypropylene
DisposableSingle-use
PackagingBlister Pack
SterilitySterile
Product LineFisherbrand
GraduatedYes
Closure TypeSnap Cap
Product TypeTest Tube
Unit SizeCS