Laboratory Notebooks
Actual product may vary
Laboratory Notebooks
Keep information safe and well organised with these laboratory notebooks. They are ideal for a variety of applications including recording research data, experimental procedures, design and development ideas.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCover ColorRuledNo. of Pages
FISFB14127569GreenGrid100
FISFB14127564BlueYes200
Catalog number FISFB14127569
Price (SGD)
303.00
Each
Cover Color:
Green
Ruled:
Grid
No. of Pages:
100
Price (SGD)
303.00
Each
  • Archival-quality acid-free pages are numbered and include spaces for title, project number, book number, date, and witness signature.
  • Lies flat when opened.
  • User data, notebook instructions, table of contents, periodic table and conversion tables are included in every book.
  • Bookmark ribbon holds your place for quick reference.
  • Covers for all versions are waterproof and chemical-resistant.
  • Lined notebooks feature college-ruled pages for easy report writing.
  • All notebooks contain periodic and conversion tables in the back.
  • Marker ribbon.
  • 8-1/2 x 11 in. (218 x 281 mm)
Note: The line model notebooks have a slight impression of vertical lines. The lines are faint -- only printed at 30%. These vertical lines are helpful if you need to add graphs or drawings to the page.
Specifications
MaterialCardboard/Paper
RuledGrid
Width (English) Page8.5 in.
Length (English) Page11 in.
ColorGreen
Cover ColorGreen
Dimensions (L x W)21.8 x 28.1 cm
Length (Metric) Page27.9 cm
Width (Metric) Page21.6 cm
No. of Pages100
Product TypeNotebook
Unit SizeEach