Polystyrene Storage Bottles
Polystyrene Storage Bottles
Actual product may vary
Polystyrene Storage Bottles
Polystyrene Storage Bottles
Fisherbrand
Polystyrene Storage Bottles
Reduce the risk of cross contamination with sterile, disposable polystyrene media bottles. Use with Bottle-Top Filters or as replacement/extra receptacles for Filter Units. Excellent as single-use storage containers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FISFB12566515500 mL
FISFB12566514250 mL
Catalog number FISFB12566515
Price (SGD)
132.00
CS
Add to cart
Volume (Metric):
500 mL
Price (SGD)
132.00
CS
Add to cart
  • Rounded with molded in hand grips
  • Polystyrene
  • Molded in graduations
  • Sterile
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionRounded with molded in hand grips
MaterialPolystyrene
SterilitySterile
TypeReceiver Flask
Volume (Metric)500 mL
Unit SizeCS