Lab Grade Cylindrical Dewar Flasks with Plastic Mesh Casing
Lab Grade Cylindrical Dewar Flasks with Plastic Mesh Casing
Actual product may vary
Lab Grade Cylindrical Dewar Flasks with Plastic Mesh Casing
Lab Grade Cylindrical Dewar Flasks with Plastic Mesh Casing
Lab Grade Cylindrical Dewar Flasks with Plastic Mesh Casing
Fisher Catalogue
Lab Grade Cylindrical Dewar Flasks with Plastic Mesh Casing
Borosilicate glass with silvered double walls and mesh casing for better grip
Have Questions?
Catalog NumberCapacity (Metric)
FISFB10197A665 mL
Catalog number FISFB10197A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
665 mL
  • Cylindrical- Wide Mouth
  • Borosilicate Glass
  • Silvered Double Walls
  • PE Stopper Included
  • For Use with Liquified Gases
  • Aluminum Housing
  • Lab Grade
  • Plastic Mesh Casing
Specifications
ColorSilver
For Use With (Application)Liquid Nitrogen
Height (Metric)265 mm
IncludesPlastic Mesh Casing
Capacity (Metric)665 mL
Depth (Metric) Interior195 mm
Diameter (English) Inner2.75 in.
Diameter (Metric) Inner70 mm
MaterialAluminum
Unit SizeEA