Lab Grade Cylindrical Dewar Flask with Aluminum Housing and Handle
Lab Grade Cylindrical Dewar Flask with Aluminum Housing and Handle
Actual product may vary
Lab Grade Cylindrical Dewar Flask with Aluminum Housing and Handle
Lab Grade Cylindrical Dewar Flask with Aluminum Housing and Handle
Lab Grade Cylindrical Dewar Flask with Aluminum Housing and Handle
Fisher Catalogue
Lab Grade Cylindrical Dewar Flask with Aluminum Housing and Handle
Borosilicate glass with silvered double walls and a convenient handle
Have Questions?
Catalog NumberCapacity (Metric)
FISFB10196104300 mL
Catalog number FISFB1019610
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
4300 mL
  • Cylindrical- Wide Mouth
  • Borosilicate Glass
  • Silvered Double Walls
  • PE Stopper Included
  • For Use with Liquified Gases
  • Aluminum Housing
  • Lab Grade
  • Coated Metal Handle
Specifications
ColorSilver
For Use With (Application)Liquid Nitrogen
Height (Metric)376 mm
IncludesHandle
Capacity (Metric)4300 mL
Depth (Metric) Interior305 mm
Diameter (English) Inner5.63 in.
Diameter (Metric) Inner143 mm
MaterialAluminum
Unit SizeEA