Reusable Glass Heavy-Duty Low-Form Beakers
Reusable Glass Heavy-Duty Low-Form Beakers
Actual product may vary
Reusable Glass Heavy-Duty Low-Form Beakers
Reusable Glass Heavy-Duty Low-Form Beakers
Reusable Glass Heavy-Duty Low-Form Beakers
Spout designed for optimum performance
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISFB101600600 mL
FISFB101250250 mL
FISFB101400400 mL
FISFB10110001000 mL
FISFB10120002000 mL
FISFB10140004000 mL
Catalog number FISFB101600
Price (SGD)
193.00
PK
Add to cart
Capacity (Metric):
600 mL
Price (SGD)
193.00
PK
Add to cart
Fisherbrand™ Reusable Glass Heavy-Duty Low-Form Beakers feature a sturdy construction ideal for mechanical shock and other harsh applications. Spout is designed for optimum performance.
  • Thick uniform walls
  • Evenly tooled top rim with extra wall thickness
  • Durable matte finish marking area
  • White graduated scale
Specifications
Accuracy±5%
Capacity (English)20.28 oz.
DescriptionBeaker
Diameter (English) Outer3.42 in.
Diameter (Metric) Outer8.7 cm
Capacity (Metric)600 mL
MaterialBorosilicate Glass
For Use With (Application)Offer Superior Mechanical Strength and Durability, Provides Improved Safety When Used Under Extreme Conditions Such as Mechanized Washing and Rough Handling
Graduation Range50 to 500 mL
Height (English)4.08 in.
Height (Metric)12.2 cm
IncludesSpout
Increments50 mL
Label TypeMatte Finish Marking
Quantity6 Pk.
TypeBeaker
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960, ASTM E438, USP Type I
ScaleDual Scale
Unit SizePK