Translucent White Wash Bottles
Translucent White Wash Bottles
Actual product may vary
Translucent White Wash Bottles
Versatile bottles with single drop or fine stream options.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISFB0340922D1000 mL
FISFB0340922C500 mL
FIS03-409-22D1000 mL
Catalog number FISFB0340922D
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
  • LDPE with one-piece polypropylene screw cap and nozzle
  • Tip may be cut back to increase flow
Specifications
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorTranslucent
MaterialLDPE
AutoclavableNot Autoclavable
Closure MaterialPolypropylene
Closure Size38 mm
Closure TypeScrew Cap
Unit SizePK