Clear Straight Sided Polystyrene Jars with White Polypropylene Caps
Clear Straight Sided Polystyrene Jars with White Polypropylene Caps
Actual product may vary
Clear Straight Sided Polystyrene Jars with White Polypropylene Caps
Wide mouth design is great for large solid samples and allow for complete removal of contents
Have Questions?
Catalog NumberCapacity (Metric)
FISFB0291226760 mL
Catalog number FISFB02912267
Price (SGD)
119.00
CS
Add to cart
Capacity (Metric):
60 mL
Price (SGD)
119.00
CS
Add to cart
  • Features wide neck and straight sides to facilitate easy recovery of contents
  • Rigid clear polystyrene jars offer glass-like clarity with a slip-resistant smooth surface
Specifications
Capacity (Metric)60 mL
Capacity (English)2 oz.
ColorClear
Diameter (Metric) Outer Neck50 mm
Diameter (Metric) Inner Neck48 mm
For Use With (Application)Large Solid Samples and Allow for Complete Removal of Contents
IncludesCap
Height (Metric) With Closure49 mm
MaterialPolystyrene
Quantity48
Closure ColorWhite
Neck Finish53-400
ClarityClear
Closure IncludedYes
MouthWide
ShapeRound
Height (Metric)46 mm
Product TypeStraight-sided Jar
Closure LinerPolyethylene Foam
Closure MaterialPolypropylene
Closure Size53-400 mm
Diameter (Metric)49 mm
Closure TypeCap
Unit SizeCS