White polypropylene screw caps provides clear surface for writing on cap
Have Questions?
Catalog Number
Neck Finish
Liner Material
Quantity
FISFB02912070
28-400
PTFE/Polyethylene Foam
100/Cs.
Catalog number FISFB02912070
Price (SGD)
205.00
PK
Add to cart
Neck Finish:
28-400
Liner Material:
PTFE/Polyethylene Foam
Quantity:
100/Cs.
Price (SGD)
205.00
PK
Add to cart
Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products