Amber Wide Mouth Straight Sided Glass Jars with White Polypropylene Caps
Amber Wide Mouth Straight Sided Glass Jars with White Polypropylene Caps
Actual product may vary
Fisherbrand
Amber Wide Mouth Straight Sided Glass Jars with White Polypropylene Caps
Amber USP Type III soda-lime glass wide mouth design is great for large solid samples and allow for complete removal of contents. Ideal for soil sampling and environmental applications
Have Questions?
Catalog NumberCapacity (Metric)
FISFB02912045120 mL
Catalog number FISFB02912045
Price (SGD)
263.00
CS
Add to cart
Capacity (Metric):
120 mL
Price (SGD)
263.00
CS
Add to cart
Designed without shoulders for maximum storage capacity. Wide mouth for easy filling
Specifications
Capacity (Metric)120 mL
Capacity (English)4 oz.
ColorAmber
Diameter (Metric) Outer Neck54 mm
Diameter (Metric) Inner Neck40 mm
For Use With (Application)Soil Sampling and Environmental Applications
IncludesCap
MaterialGlass
Quantity24
Closure ColorWhite
Neck Finish58-400
Certifications/ComplianceUSP Type III Soda Lime Glass Wide Mouth Design is Great for Large Solid Samples and Allow for Complete Removal of Contents
Closure IncludedYes
MouthWide
ShapeRound
Height (Metric)69 mm
Product TypeStraight-sided Jar
Closure LinerPTFE/Polyethylene Foam
Closure MaterialPolypropylene
Closure Size58-400 mm
Diameter (Metric)60 mm
Closure TypeCap
Unit SizeCS