Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Closure Liner | Quantity |
---|---|---|---|
FISFB02911789 | 240 mL | PTFE/Polyethylene Foam | 12 |
FBWW014020 | 60 mL | Pulp/Vinyl | 24 |
FISFB02911780 | 120 mL | Pulp/Vinyl | 24 |
FISFB02911753 | 480 mL | PTFE/Polyethylene Foam | 12 |
FISFB02911754 | 950 mL | PTFE/Polyethylene Foam | 12 |