Clear Boston Round Glass Bottles with Black Phenolic Caps
Clear Boston Round Glass Bottles with Black Phenolic Caps
Actual product may vary
Clear Boston Round Glass Bottles with Black Phenolic Caps
Clear Boston round Type III soda-lime glass is ideal for solvent, chemical or sample storage. Packaged in convenience packs with black phenolic caps attached to maintain cleanliness
Have Questions?
Catalog NumberCapacity (Metric)Closure Liner
FISFB0291178230 mLPolyethylene Cone
Catalog number FISFB02911782
Price (SGD)
132.00
CS
Add to cart
Capacity (Metric):
30 mL
Closure Liner:
Polyethylene Cone
Price (SGD)
132.00
CS
Add to cart
Clear Boston round Type III soda-lime glass is ideal for solvent, chemical or sample storage. Packaged in convenience packs with black phenolic caps attached to maintain cleanliness
  • General use bottles ideal for when content integrity and sample or product visibility is important
  • Narrow screw neck opening, curved shoulders, round body
  • Glass offers minimal chemical interference with contents
  • Clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary
  • Environmentally sensitive - help eliminate waste and insure product integrity for long term storage
Specifications
Capacity (Metric)30 mL
Capacity (English)1 oz.
ColorClear
Diameter (Metric) Outer Neck18 mm
Diameter (Metric) Inner Neck10 mm
Height (Metric) With Closure81 mm
MaterialGlass (Type 3)
Quantity48
Closure ColorBlack
Neck Finish20-400
Closure IncludedYes
MouthNarrow
Height (Metric)79 mm
Product TypeBoston Round Bottle
Closure LinerPolyethylene Cone
Closure MaterialPhenolic
Closure Size20-400 mm
Diameter (Metric)31 mm
Unit SizeCS