Amber Boston Round Glass Bottles with White Polypropylene Caps
Amber Boston Round Glass Bottles with White Polypropylene Caps
Actual product may vary
Amber Boston Round Glass Bottles with White Polypropylene Caps
Amber Boston round Type III soda-lime glass is ideal for solvent, chemical or sample storage. Packaged in convenience packs with white polypropylene caps attached to maintain cleanliness.
Have Questions?
Catalog NumberCapacity (Metric)Closure Liner
FISFB02911741240 mLPTFE/Polyethylene Foam
Catalog number FISFB02911741
Price (SGD)
52.00
CS
Add to cart
Capacity (Metric):
240 mL
Closure Liner:
PTFE/Polyethylene Foam
Price (SGD)
52.00
CS
Add to cart
Amber Boston round Type III soda-lime glass is ideal for solvent, chemical or sample storage. Packaged in convenience packs with white polypropylene caps attached to maintain cleanliness
  • General use bottles ideal for when content integrity and UV protection is important
  • Feature a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorbs all wavelengths in the ultraviolet spectrum, ensures integrity for long term storage, and offers excellent protection for UV-sensitive contents
  • White Polypropylene cap offers good impact strength, thermal resistance and pliability
Specifications
Capacity (Metric)240 mL
Capacity (English)8 oz.
ColorAmber
Diameter (Metric) Outer Neck21 mm
Diameter (Metric) Inner Neck13 mm
Height (Metric) With Closure139 mm
MaterialGlass (Type 3)
Quantity12
Closure ColorWhite
Neck Finish24-400
Closure IncludedYes
MouthNarrow
Height (Metric)136 mm
Product TypeBoston Round Bottle
Closure LinerPTFE/Polyethylene Foam
Closure MaterialPolypropylene
Closure Size24-400 mm
Diameter (Metric)60 mm
Unit SizeCS