Clear Graduated Glass Bottles with White Polypropylene Cap
Clear Graduated Glass Bottles with White Polypropylene Cap
Actual product may vary
Clear Graduated Glass Bottles with White Polypropylene Cap
Fisherbrand™ Clear Graduated Glass Bottles with White Polypropylene Cap feature a wide mouth for easy removal of contents.
Have Questions?
Catalog NumberCapacity (Metric)
FISFB02911460A120 mL
Catalog number FISFB02911460A
Price (SGD)
130.00
CS
Add to cart
Capacity (Metric):
120 mL
Price (SGD)
130.00
CS
Add to cart
  • Graduated in both milliliters and ounces
  • Clear glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
Specifications
Capacity (Metric)120 mL
Capacity (English)4 oz.
ColorClear
Diameter (Metric) Outer Neck45 mm
Diameter (Metric) Inner Neck35 mm
Height (Metric) With Closure105 mm
MaterialGlass (Type 3)
Quantity24
Closure ColorWhite
Neck Finish48-400
Closure IncludedYes
GraduatedYes
MouthWide
Height (Metric)102 mm
Product TypeGraduated Bottle
Closure LinerPTFE/Polyethylene Foam
Closure MaterialPolypropylene
Closure Size48-400 mm
Diameter (Metric)51 mm
Unit SizeCS