Search Fisher Scientific
| Catalog Number | Capacity (Metric) | Closure Liner |
|---|---|---|
| FISFB02911443 | 950 mL | PTFE/Polyethylene Foam |
| FISFB02911438 | 30 mL | PTFE/Polyethylene Foam |
| FISFB02911440 | 120 mL | PTFE/Polyethylene Foam |
| FISFB02911439 | 60 mL | PTFE/Polyethylene Foam |
| FISFB02911441 | 250 mL | PTFE/Polyethylene Foam |