Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) |
---|---|
FISFB012919 | 30 mL |
FISFB012918 | 15 mL |
FISFB0129110 | 50 mL |
FISFB012917 | 100 mL |
FISFB0129111 | 150 mL |
FISFB012915 | 250 mL |
FISFB0129112 | 400 mL |
FISFB012916 | 600 mL |