Ultraviolet Viewing Cabinets
Actual product may vary
Spectroline
Ultraviolet Viewing Cabinets
Designed for viewing electrophoresis gels and TLC plates
Have Questions?
Catalog NumberSizeUV Monitoring
FISFB-PDS-81MediumYes
Catalog number FISFB-PDS-81
Price (SGD)
-
Size:
Medium
UV Monitoring:
Yes
  • Cabinet housing is sturdy aluminum with chemical-resistant polyurethane finish
  • Removable bottom panel allows units to be placed underneath cabinet for higher UV intensity and greater fluorescent contrast
  • Cabinets can be converted into benchtop photodocumentation systems using any 35mm SLR camera; snap-on adapters can be ordered through your Fisher Scientific Customer Service Representative or by visiting www.fishersci.com
  • Small Cabinets have space to view two 20 × 20cm gels or plates; can accommodate two double-tube UV lamps
  • Large Cabinets have space for four 20 × 20cm gels or plates; accept two larger lamps
  • Small cabinet housing dimensions: 17 1/2L × 20 1/4W × 9 3/4 in.H (45 × 51 × 25cm)
  • Large cabinet housing dimensions: 19 1/2L × 24 1/4W × 12 3/4 in.H (50 × 62 × 32cm)
Specifications
Height (English)9.75 in.
Width (English)20.25 in.
IncludesBuilt-in 25w white light source
Height (Metric)24.7cm
Width (Metric)51.4cm
UV MonitoringYes
SizeMedium
Product TypeUltraviolet Viewing Cabinet Transilluminator
Unit SizeEach