PIPETMANTM TIPACKTM Racked Pipet Tips
Actual product may vary
PIPETMANTM TIPACKTM Racked Pipet Tips
PIPETMANTM TIPACKTM Racked Pipet Tips
Gilson
PIPETMANTM TIPACKTM Racked Pipet Tips
Engineered to be functional, convenient and robust. GilsonTM PIPETMANTM TIPACKTM Racked Pipet Tips are available in sterilized or autoclavable packages.
Have Questions?
Catalog NumberLength (Metric)Volume (Metric)ModelSterility
FISF171301G14.5 cm2 to 200 μLD200Sterile
Catalog number FISF171301G
Price (SGD)
259.00
Pack
Length (Metric):
14.5 cm
Volume (Metric):
2 to 200 μL
Model:
D200
Sterility:
Sterile
Price (SGD)
259.00
Pack
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Requires low tip-mounting force.
  • Robust and designed to withstand autoclaving.
  • Sterility assurance level SAL 10-6 guaranteed, using gamma irradiation.
  • All packs are wrapped in medical device quality plastic which provides a complete shield against contamination.
  • Indicator dots used on Gilson products undergo an irreversible color change upon sterilization; yellow gamma dots turn red when products have been properly exposed.
  • TIPACKs are securely sealed with a sticker; a full product description is given on the TIPACK for traceability and easy identification.
  • Available in sterilized or autoclavable packages.
  • Manufactured in a clean room and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Easy identification with color-coded packaging to meet GLP requirements.
Specifications
Volume (Metric)2 to 200 μL
FilteredNon-filtered
SterilitySterile
ColorClear
AutoclavableAutoclavable
ModelD200
Length (English)5.71 in.
Length (Metric)14.5 cm
Clean ClaimsDNA free, RNA free, RNAse free, DNAse free, ATP free, Endotoxin free
FormatRack
Tip StyleStandard
Quantity960 (10x96)
Product TypePipet Tips
DisposableSingle-use
For Use With (Equipment)Universal Fit
GraduatedYes
Product LinePipetman Tipack
MaterialPolypropylene
Graduations20 μL; 100 μL
Unit SizePack