DISTRITIPSTM Syringes
DISTRITIPSTM Syringes
Actual product may vary
Fisherbrand
DISTRITIPSTM Syringes
Guarantees accuracy and precision. DISTRITIPSTM Syringes have an aliquot volume range from 1 μL to 1.25 mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Model
FISF164150G100 μL to 1.25 mL12.5 mL Maxi ST (Sterile)
FISF164140G10 to 125 μL1250 μL Mini ST (Sterile)
Catalog number FISF164150G
Price (SGD)
483.00
PK
Add to cart
Volume (Metric):
100 μL to 1.25 mL
Model:
12.5 mL Maxi ST (Sterile)
Price (SGD)
483.00
PK
Add to cart
  • With level indicator
  • Available in standard or sterilized (ST) form, in boxes of 50
  • Certified free of RNase, DNase, DNA, RNA, and Protease
  • Free of dyes, so no risk of contamination from their components
  • Only three syringes are needed to cover 1 μL to 1.25 mL
  • More than 100 aliquots can be dispensed from each syringe
  • Ideal for precise aliquots
  • Can be used with viscous and volatile solutions
  • Bulk or Individually wrapped
Specifications
Volume (Metric)100 μL to 1.25 mL
FilteredNon-filtered
SterilitySterile
ColorClear
AutoclavableAutoclavable
Model12.5 mL Maxi ST (Sterile)
Accuracy±10.0 μL
Length (English)4.4 in.
Length (Metric)112 mm
Clean ClaimsFree of DNA, RNA, RNase, DNase, Pyrogens
For Use With (Application)Viscous, volatile solutions
Sterilization MethodGamma Irradiated
CompatibilityDistriman
FormatIndividually Wrapped
Tip StylePositive Displacement
PackagingIndividually Wrapped
Quantity50 Pk.
Product TypeDistriTip™ Syringe
DisposableSingle-use
For Use With (Equipment)DISTRIMAN Repeating Pipette
GraduatedYes
Product LineDistriTips