MICROMANTM E Capillary Pistons
MICROMANTM E Capillary Pistons
Actual product may vary
MICROMANTM E Capillary Pistons
MICROMANTM E Capillary Pistons
MICROMANTM E Capillary Pistons
MICROMANTM E Capillary Pistons provide reproducible, reliable results even with the most viscous or volatile liquids.
Have Questions?
Catalog NumberQuantityVolume (Metric)ModelFormat
FISF148414G192 Pk.10 to 100 μLCP100Box
Catalog number FISF148414G
Price (SGD)
489.00
PK
Add to cart
Quantity:
192 Pk.
Volume (Metric):
10 to 100 μL
Model:
CP100
Format:
Box
Price (SGD)
489.00
PK
Add to cart
  • Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
  • Made specifically for GilsonTM MICROMANTM E pipets.
  • Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs.
Specifications
Volume (Metric)10 to 100 μL
FilteredNon-filtered
SterilityNon-sterile
ColorClear
AutoclavableAutoclavable
ModelCP100
Length (English)4.8 in.
Length (Metric)41.8 mm
CompatibilityMicroman E
FormatBox
Tip StylePositive Displacement
PackagingBox
Quantity192 Pk.
Product TypeCapillary Piston
DisposableSingle-use
For Use With (Equipment)Gilson Microman
MaterialPolypropylene
Fit for PurposeCertified Quality
Unit SizePK