▸
Biochemical Reagents
▸
Staining Reagents
▸
Stains and Dyes
▸
Lactophenol Cotton Blue Stain, Certified
Actual product may vary
Eng Scientific
Lactophenol Cotton Blue Stain, Certified
For staining fungi using Lactophenol Cotton Blue Stain.
Have Questions?
Catalog Number
Quantity
FISES6731
1 x 100 mL
Catalog number
FISES6731
Price (SGD)
-
Quantity:
1 x 100 mL
Lactic acid acts as a preservative for fungi, killed by phenol and stained by cotton blue
Fungal elements are stained a deep blue; background is pale blue
Lactophenol Cotton Blue Stain
Includes Phenol, lactic acid, glycerol, aniline blue
For use with staining fungi, in-vitro diagnostic use/li>
For in vitro diagnostic use only.
Specifications
Type
Lactophenol Cotton Blue Stain
Quantity
1 x 100 mL
Includes
Phenol, lactic acid, glycerol, aniline blue
Format
Bottle
For Use With (Application)
Staining fungi, in-vitro diagnostic use
Stain Type
Lactophenol Cotton Blue