Deepwell Plates 96-well, 2000 μL
Deepwell Plates 96-well, 2000 μL
Actual product may vary
Deepwell Plates 96-well, 2000 μL
Deepwell Plates 96-well, 2000 μL
Deepwell Plates 96-well, 2000 μL
Deepwell Plates 96-well, 2000 μL
Fisherbrand
Deepwell Plates 96-well, 2000 μL
Eppendorf Deepwell plates offer efficient sample handling with features like OptiTrack™ matrix for rapid identification and g-Safe™ technology for stability during centrifugation. SafeCode variants provide secure three-level coding, including error correction for reliable identification, ensuring flexibility and accuracy in experimental workflows.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorPurity or Quality GradePackaging
FISE951033600White, WhitePCR Clean (Non-sterile)10 Bags x 8 Plates
FISE951033405White, WhitePCR Clean (Non-sterile)5 Bags x 4 Plates
FIS13-864-302BluePCR Clean (Non-sterile)5 Bags x 4 Plates
Catalog number FISE951033600
Price (SGD)
2,209.00
CS
Add to cart
Color:
White, White
Purity or Quality Grade:
PCR Clean (Non-sterile)
Packaging:
10 Bags x 8 Plates
Price (SGD)
2,209.00
CS
Add to cart
  • OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling.
  • RecoverMax™ well design: Optimized well geometry for maximum sample recovery and excellent mixing properties.
  • g-Safe™: exceptional centrifugation stability up to 6,000 x g for faster protocols and improved sample quality.
  • Raised well rims and smooth surface for reliable closing, even with heat sealing.
  • Easy and reliable stacking.
  • Minimized remaining volume and high uniformity from well to well.
  • High-quality polypropylene ensures high resistance to chemicals, mechanical stress and temperature extremes.
  • Manufactured without slip agents, plasticizers, and biocides – Materials which could have a negative effect on bioassays.
  • Autoclavable (121°C, 20 min.).
  • Safe Code option provides 3-level coding for safe sample and vessel identification.
  • ECC 200 error correction function enables safe code reading with up to 30% damaged codes.
Specifications
BarcodeNo Barcode
Certifications/ComplianceIVD, SBS/ANSI
FormatStandard
LidWithout Lid
Array8 x 12
AutoclavableAutoclavable
Packaging10 Bags x 8 Plates
ColorWhite, White
Product TypeDeepwell Microplate
MaterialPolypropylene
Volume (Metric) Well2000 μL
No. of Wells96
Volume (Metric) Working50 to 2,000 μL
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeRound Bottom (U-well)
Dnase FreeYes
For Use With (Application)Ideal for sample preparation or long-term storage of nucleic acids in forensic, microarrays, next generation sequencing applications
Max. RCF6000 x g
Purity or Quality GradePCR Clean (Non-sterile)
Quantity80 Pk.
RNase FreeYes
Temperature Range (English)-122.8°F to 212°F
Temperature Range (Metric)-86°C to +100°C
Well ColorColorless
Dimensions (L x W x H)85.5 x 127.8 x 44.1 mm
Width (Metric)12.78 cm
Height (Metric)4.41 cm
Depth (Metric)8.55 cm
Unit SizeCS