twin.tec™ PCR Plates 96-well, 150 μL - PCR Plates
twin.tec™ PCR Plates 96-well, 150 μL - PCR Plates
Actual product may vary
Fisherbrand
twin.tec™ PCR Plates 96-well, 150 μL - PCR Plates
Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. Raised well rims enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSkirt StyleProduct TypePackaging
FISE951020401ColorlessSkirtedStandard PCR Plate5 Bags x 5 Plates
FISE9-510-20427YellowSkirtedStandard PCR Plate5 Bags x 5 Plates
FISE951020460BlueSkirtedStandard PCR Plate5 Bags x 5 Plates
Catalog number FISE951020401
Price (SGD)
556.00
CS
Add to cart
Color:
Colorless
Skirt Style:
Skirted
Product Type:
Standard PCR Plate
Packaging:
5 Bags x 5 Plates
Price (SGD)
556.00
CS
Add to cart
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
Specifications
BarcodeNo Barcode
Certifications/ComplianceHuman DNA, DNase, RNase-free
FormatStandard
Array8 x 12
AutoclavableAutoclavable
Packaging5 Bags x 5 Plates
ColorColorless
Product TypeStandard PCR Plate
Skirt StyleSkirted
MaterialPolypropylene, Polycarbonate
Volume (Metric) Well150 μL
No. of Wells96
SterilityNon-sterile
For Use With (Application)Ideal for extreme applications such as multiple heat sealing.
Max. RCF2250 x g
Purity or Quality GradePCR Clean (Non-sterile)
Quantity25 Pk.
Well ColorColorless
Unit SizeCS