Protein LoBind™ Tubes
Protein LoBind™ Tubes
Actual product may vary
Fisher Catalogue
Protein LoBind™ Tubes
Engineered to minimize sample binding and elevate recovery, Eppendorf™ Protein LoBind™ Tubes safeguard your valuable proteins during processing. With cutting-edge LoBind technology, ensure optimal yields for your Western blotting, mass spectrometry, and protein purification applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISE9250000901.5 mL
FIS13-698-7941.5 mL
FIS13-698-7930.5 mL
FIS13-698-7952 mL
Catalog number FISE925000090
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1.5 mL
  • Surface-coating-free, including substances like silicone, to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Snap, screw, and SnapTec™ cap options available.
Specifications
Certifications/ComplianceDNA, DNase, RNase, PCR Inhibitors free
Capacity (Metric)1.5 mL
ColorClear
MaterialPolypropylene
For Use With (Application)Protein LoBind consumables are best suited for work in protein research, mass spectrometry, protein array sample preparation, and storage of peptide, antibody or virus samples
Packaging400
SterilityNon-sterile
Closure TypeSafe-lock Lid
Bottom ShapeConical
Max. RCF30,000 x g
Unit SizePK