UVette™ Cuvets
UVette™ Cuvets
Actual product may vary
UVette™ Cuvets
UVette™ Cuvets
UVette™ Cuvets
Fisherbrand
UVette™ Cuvets
An alternative to the expensive, breakable quartz cuvets traditionally required for use at UV wavelengths
Have Questions?
Catalog NumberCapacity (Metric)Description
FISE409910000850 μLUVette Routine Pack: 200 cuvets in reclosable box that supports routine use
Catalog number FISE4099100008
Price (SGD)
955.00
EA
Add to cart
Capacity (Metric):
50 μL
Description:
UVette Routine Pack: 200 cuvets in reclosable box that supports routine use
Price (SGD)
955.00
EA
Add to cart
  • UV- and VIS-transparent between 220 and 1600 nm
  • Suitable for measuring small volumes (≥50 μL)
  • DNase-, RNase-, and protein-free
  • Disposable
  • Measure samples with a normal concentration using the 10 mm optical pathlength; for samples with a higher concentration, rotate the UVette cuvets by 90° and use the shorter 2 mm optical pathlength—no need to dilute sample
  • UVette Cuvets: 80 original Eppendorf disposables, single sealed cuvettes
  • UVette Starter Kit: 80 UVettes + 1 universal adapter for a 15 mm light center height (including GeneQuant); convertible to 8.5 mm
  • UVette Routine Pack: Eppendorf Quality purity level, reclosable box of 200 pcs.
Specifications
TypeUvette Routine Pack
DescriptionUVette Routine Pack: 200 cuvets in reclosable box that supports routine use
DisposableSingle-use
Capacity (Metric)50 μL
MaterialUV-transparent Plastic
IncludesEppendorf quality purity level, reclosable box of 200 pieces
Quantity1 Ea.
Path Length10 and 2 mm
Wavelength Range220 to 1600 nm
For Use With (Equipment)Biophotometer, Spectrophotometer
For Use With (Application)UV/Vis Spectroscopy
Unit SizeEA