Heat Sealer Film and Foils
Heat Sealer Film and Foils
Actual product may vary
Fisherbrand
Heat Sealer Film and Foils
Create 100% hermetic seal, eliminate evaporation, and prevent cross-contamination
Have Questions?
Catalog NumberProduct Type
FISE0030127854PCR Foil
Catalog number FISE0030127854
Price (SGD)
705.00
PK
Add to cart
Product Type:
PCR Foil
Price (SGD)
705.00
PK
Add to cart
  • Requires Eppendorf heat sealer (Cat. No. E5390 000 016)
  • Transparent heat sealing film allows sample monitoring in the plates
  • Aluminum coated, heat-sealing foils are available in peel-off or pierceable versions
  • Foil stripper tool removes heat-sealed film and foils completely
  • Certified free of DNA, DNase, RNase, and PCR inhibitors
  • Autoclavable
Specifications
MaterialAluminum, Polypropylene
AutoclavableAutoclavable
No. per Pack100 Pk.
For Use With (Application)Heat sealing PCR foil for sealing plates, especially recommended for low reaction volumes, best protection against evaporation
For Use With (Equipment)Automated systems
Product TypePCR Foil
Certified RNase/DNase-freeYes
Temperature Range (Metric)-80°C to +100°C
Unit SizePK