PCR Tubes - Singles and Strips
Actual product may vary
Fisherbrand
PCR Tubes - Singles and Strips
Eppendorf PCR Tubes ensure uniform heat distribution to the sample for optimal PCR performance. The user-friendly design allows for easy opening and provides a secure seal to prevent evaporation.
Have Questions?
Catalog NumberClosure TypeFormatPurity or Quality GradeVolume (Metric)
FISE0030124286Hinged Cap8-tube StripsPCR Clean (Non-sterile)0.2 mL
Catalog number FISE0030124286
Price (SGD)
1,065.00
Case
Closure Type:
Hinged Cap
Format:
8-tube Strips
Purity or Quality Grade:
PCR Clean (Non-sterile)
Volume (Metric):
0.2 mL
Price (SGD)
1,065.00
Case
Ensure efficient and homogenous heat transfer with these thin-walled polypropylene tubes. Eppendorf™ Polypropylene PCR Tubes are easy to open, but provide tight sealing to prevent evaporation in PCR.
  • Design ensures an airtight seal to prevent contamination and evaboration.
  • Thin polypropylene walls ensure efficient and homogeneous heat transfer or fast and high-quality PCR results.
  • 8 reaction tubes in strip-format – ideal for small sample volumes. Easily adaptable for automation.
  • Strips are sealable using flat or domed strip-lids.
  • PCR Clean: Certified free from human DNA, DNase, RNase and PCR inhibitors).
Specifications
Certifications/ComplianceHuman DNA, DNase, RNase, PCR inhibitor-free
ColorClear
Volume (Metric)0.2 mL
Quantity120 Pk.
For Use With (Equipment)0.2mL thermal cycler blocks for efficient and homogenous heat transfer to the sample
SterilityNon-sterile
MaterialPolypropylene
Purity or Quality GradePCR Clean (Non-sterile)
Closure TypeHinged Cap
AutoclavableAutoclavable
Format8-tube Strips
Product TypeThin Wall PCR Tubes (8-tube Strip )
Unit SizeCase