MAbPac™ SCX-10 HPLC Columns
MAbPac™ SCX-10 HPLC Columns
Actual product may vary
Thermo Scientific
MAbPac™ SCX-10 HPLC Columns
Use these strong cation-exchange columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)Particle SizeColumn Format
FISDX07463150 mm4 mm10 μmGuard Column
Catalog number FISDX074631
Price (SGD)
2,154.00
EA
Add to cart
Length (Metric):
50 mm
Diameter (Metric):
4 mm
Particle Size:
10 μm
Column Format:
Guard Column
Price (SGD)
2,154.00
EA
Add to cart
Use Thermo Scientific™ MAbPac™ SCX-10 LC Columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants. The unique nonporous pellicular resin provides the resolving power to separate mAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.
Superior Performance for mAb Variant Analysis
  • Exceptionally high resolution for mAb variants
  • Ideal for characterization and quality control assessment of mAbs
  • Well-suited for stability studies, mAb therapeutic method development and high-throughput screening
  • Unmatched column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
  • Meets the regulatory requirements for biopharmaceutical characterization
Multiple Sizes Available
  • Available in 3, 5, and 10μm particle sizes in standard and UHPLC formats
  • Smaller-particle-size columns provide the same resolution as the 10μm columns, but with significantly faster analysis time
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatGuard Column
Diameter (Metric)4 mm
Length (Metric)50 mm
Max. Pressure3000 psi (207 bar)
Particle Size10 μm
pH2 to 12
Pore SizeNonporous
QuantityEach
TemperatureAmbient to 60°C
Stationary PhaseSulfonate
Product LineMAbPac
Packing MaterialSpherical, Nonporous Polymer
Column TypeIon Exchange
PhaseIon Exchange
TypeGuard Column
TechniqueBio-LC
Green FeaturesResponsibly packaged
Unit SizeEA