DNAPac™ PA200 Oligonucleotide HPLC Columns
DNAPac™ PA200 Oligonucleotide HPLC Columns
Actual product may vary
Thermo Scientific
DNAPac™ PA200 Oligonucleotide HPLC Columns
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
FISDX063000250 mm4 mm8 μm
FISDX06299850 mm4 mm8 μm
Catalog number FISDX063000
Price (SGD)
6,870.00
EA
Add to cart
Length (Metric):
250 mm
Diameter (Metric):
4 mm
Particle Size:
8 μm
Price (SGD)
6,870.00
EA
Add to cart
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
  • Achieve n, n-1 resolution for oligonucleotides
  • Resolve oligonucleotides with secondary structures
  • Assay phosphorothioate purity
  • Selectivity control with eluent pH, salt, and solvent
  • Resolve RNA with aberrant (2' – 5') links from normal SSRNA
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4 mm
Flow Rate1.2 mL/min
For Use WithUltiMate 3000 Titanium System
Length (Metric)250 mm
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 2 to 12.5, 100% compatible with common organic solvents, Ionic form eluents: chloride, perchlorate
Particle Size8 μm
pH2.5 to 12.5
Pore SizeNonporous
QuantityEach
Temperature85°C
Stationary PhaseQuaternary Ammonium
Product LineDNAPac
Packing MaterialSpherical, Nonporous Polymer
Column TypeIon Exchange
PhaseIon Exchange
TypeAnalytical Column
TechniqueBio-LC
Green FeaturesResponsibly packaged
Unit SizeEA