Cell-VU Prestained Slides
Cell-VU Prestained Slides
Actual product may vary
Fisherbrand
Cell-VU Prestained Slides
Used for simplified and convenient manual hematology staining that is safer with less exposure to reagents and stains
Have Questions?
Catalog number FISDRM900
Price (SGD)
-
Used for simplified and convenient manual hematology staining that is safer with less exposure to reagents and stains
  • CELL-VU™ pre-stained Hematology/Morphology slides combine conventional microscope slides with a combination of methylene blue and cresyl violet stains into a single, ready to use device.
  • Samples are applied directly to the slide and a coverslip is placed over the sample. One step process limits exposure to reagents and potentially harmful stains.
  • Each slide contains a dried layer of stain that allows for rapid visualization and differential cell assessment from whole blood and body fluids such as CSF, and urine samples.
  • CELL-VU™ can also be used for World Health Organization and Kruger strict morphology sperm assessment from undiluted semen.
Specifications
Regulatory StatusFDA registered
Stain TypeMethylene Blue and Cresyl Violet
Length (Metric)76.2 mm
MaterialGlass
TypePre-Stained Morphology Slides
Width (Metric)25.4 mm
ColorBlue
Length (English)3 in.
Quantity75
Width (English)1 in.
Unit SizePK