CELL-VU™ Disposable Sperm Count Cytometers
CELL-VU™ Disposable Sperm Count Cytometers
Actual product may vary
CELL-VU™ Disposable Sperm Count Cytometers
Cytometers, Sperm Count; CELL-VU; For undiluted specimens; Disposable; 3 x 1 in. glass slide and coverslip w/counting grid etched on surface; Slide has 2 chambers for dual determinations; 25 slides and 50 cover slips/box
Have Questions?
Catalog number FISDRM600
Price (SGD)
733.00
PK
Add to cart
Price (SGD)
733.00
PK
Add to cart
Cytometers, Sperm Count; CELL-VU; For undiluted specimens; Disposable; 3 x 1 in. glass slide and coverslip w/counting grid etched on surface; Slide has 2 chambers for dual determinations; 25 slides and 50 cover slips/box
  • For sperm counts in undiluted specimens
  • The CELL-VU Cytometer consists of a standard (3 × 1 in.), specially-designed glass slide and a coverslip with the counting grid laser-etched into its surface
  • Disposal of the entire apparatus eliminates cleaning and minimizes clinician exposure to body fluids
  • Both the slide and coverslip are marked to ensure correct use
  • The coverslip's 1 × 1mm grid is divided into one-hundred 0.1mm areas and is thin enough to use under high magnification without special adapters or reticles
  • This depth is optimal for sperm cells to form in a monolayer, movement is unencumbered, motility can be assessed and counts are made easily
Specifications
Product TypeDisposable Cytometer
DisposableSingle-use
MaterialGlass
Quantity25 Pk.
Total Count2.0
Depth (English)0.0007 in.
Depth (Metric)20μm
For Use With (Application)Sperm Counts in Undiluted Specimens
Includes25 Slides and 50 coverslips
Unit SizePK