MF-Millipore™ Membrane Filter, 0.65 μm
MF-Millipore™ Membrane Filter, 0.65 μm
Actual product may vary
MilliporeSigma
MF-Millipore™ Membrane Filter, 0.65 μm
MF-Millipore™ Membrane Filter composed of mixed cellulose esters (MCE) membrane, hydrophilic.
Have Questions?
Catalog NumberDiameter (Metric)
FISDAWP0470047 mm
Catalog number FISDAWP04700
Price (SGD)
682.00
PK
Add to cart
Diameter (Metric):
47 mm
Price (SGD)
682.00
PK
Add to cart
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.
MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features and Benefits:
  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
AutoclavableNot Autoclavable
ColorWhite
DisposableReusable
Filter TypeMembrane Filter
FormatDisc
MaterialMCE
Pore Size0.65 μm
SterilityNon-sterile
Flow Rate140 mL/min. x cm2
HydrophobicityHydrophilic
TypeFilter Membranes
WettabilityHydrophilic
Diameter (English)1.9 in.
Diameter (Metric)47 mm
Unit SizePK