ITS+ Premix Universal Culture Supplement
ITS+ Premix Universal Culture Supplement
Actual product may vary
ITS+ Premix Universal Culture Supplement
Stimulates proliferation of a variety of cells under serum-reduced conditions. Corning™ ITS+ Premix Universal Culture Supplement contains insulin, human transferrin and selenous acid, the three most universally esential components of defined culture media.
Have Questions?
Catalog NumberQuantity
FISCB-4035220 mL
Catalog number FISCB-40352
Price (SGD)
-
Quantity:
20 mL
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
Specifications
Product TypeITS Premix
SterilitySterile-filtered
FormulationAs an aqueous solution containing human recombinant insulin, human transferrin (12.5 mg each), selenous acid (12.5 μg), BSA (2.5 g), and linoleic acid (10.7 mg)
Quantity20 mL
Unit SizeEA
Contents & Storage
2° to 8°C