Attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, FEP Polymer, and biological materials. Corning™ Cell-Tak Cell and Tissue Adhesive is a specially formulated protein solution extracted from marine mussel, Mytilus edulis.
Have Questions?
Change view
Catalog Number
Quantity
FISCB-40241
5 mg
FISCB-40242
10 mg
2 Options
Catalog number FISCB-40241
Price (SGD)
-
Quantity:
5 mg
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Specifications
DescriptionMolecular weight: 110 to 114kd
Concentration1 to 5μg/ cm2 of growth surface depending on cell type
FormLiquid
Product TypeCell and Tissue Adhesive
FormulationAs liquid in 5% acetic acid
Quantity5 mg
SourcePolyphenolic Proteins secreted by Mytilus edulis
Unit SizeEach
Contents & Storage
Stable for at least three months at 2° to 8°C. Do not freeze.