1x1 image pixel for data collection
Matrigel™ Matrix
Matrigel™ Matrix
Actual product may vary
Matrigel™ Matrix
Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.
Have Questions?
Catalog NumberQuantityWithout Additives
FISCB-40234A5 mLNo LDEV
Catalog number FISCB-40234A
Price (SGD)
-
Quantity:
5 mL
Without Additives:
No LDEV
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Specifications
Product TypeBasement Membrane Matrix
Without AdditivesNo LDEV
For Use With (Application)Cell growth and differentiation; metabolism/toxicology studies; invasion assays; in vitro and in vivo angiogenesis assays; in vivo angiogenesis studies and augmentation of tumors in immunosuppressed mice
Product LineMatrigel
Quantity5 mL
SourceEngelbreth-Holm-Swarm Mouse Tumor
Unit SizeEA
Contents & Storage
-20°C
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection