Matrigel™ GFR Basement Membrane Matrix
Actual product may vary
Matrigel™ GFR Basement Membrane Matrix
Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ GFR Basement Membrane Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.
Have Questions?
Catalog NumberQuantityWithout Additives
FISCB-4023010 mLNo LDEV
Catalog number FISCB-40230
Price (SGD)
-
Quantity:
10 mL
Without Additives:
No LDEV
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Specifications
Product TypeGFR Basement Membrane Matrix
Without AdditivesNo LDEV
For Use With (Application)Suitable for applications that require a highly defined basement membrane preparation.
Quantity10 mL
SourceEngelbreth-Holm-Swarm Mouse Tumor
Unit SizeEach
Contents & Storage
-20°C