Fibronectin, Human
Fibronectin, Human
Actual product may vary
Fibronectin, Human
Promotes cellular attachment, spreading, and proliferation on tissue-culture surfaces. Corning™ Fibronectin, Human can be used as an additive to serum-free culture media.
Have Questions?
Catalog NumberQuantity
FISCB-400081 mg
Catalog number FISCB-40008
Price (SGD)
-
Quantity:
1 mg
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
The principal functions of fibronectin appear to be in cellular migration during wound healing and development, regulation of cell growth and differentiation, and haemostasis/thrombosis.
Specifications
Product TypeFibronectin
pH11
Quantity1 mg
SourceHuman Plasma
Unit SizeEA
Contents & Storage
2° to 8°C