Endothelial Cell Growth Supplement (ECGS)
Endothelial Cell Growth Supplement (ECGS)
Actual product may vary
Endothelial Cell Growth Supplement (ECGS)
Ideal for endothelial cell culture
Have Questions?
Catalog NumberQuantity
FISCB-4000615 mg
Catalog number FISCB-40006
Price (SGD)
-
Quantity:
15 mg
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
Specifications
Product TypeGrowth Supplement
Quantity15 mg
Unit SizeEA
Contents & Storage
2° to 8°C