Phenolic GPI Screw Caps for Disposable Culture Tubes
Phenolic GPI Screw Caps for Disposable Culture Tubes
Actual product may vary
Phenolic GPI Screw Caps for Disposable Culture Tubes
Phenolic GPI Screw Caps for Disposable Culture Tubes
Phenolic GPI Screw Caps for Disposable Culture Tubes
With white rubber liner
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure SizeHeight (Metric)
FISC99999-1313-41513.5 mm
FIS14-962-30D15-41516 mm
Catalog number FISC99999-13
Price (SGD)
749.00
CS
Add to cart
Closure Size:
13-415
Height (Metric):
13.5 mm
Price (SGD)
749.00
CS
Add to cart
Made of a phenolic compound. These caps have a white rubber liner.
Specifications
Closure Size13-415
Liner MaterialRubber
Product TypeScrew Cap
AutoclavableAutoclavable
MaterialPhenolic
ColorBlack
Height (Metric)13.5 mm
No. per Pack1000/Pk.
No. per Case1000/Cs.
For Use With (Equipment)Culture Tubes
Unit SizeCS