Check Order Status
Calcium Chloride Dihydrate (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Calcium Chloride Dihydrate (Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FISC79-500
500 g
Poly Bottle
Catalog number
FISC79-500
Price (SGD)
-
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Calcium Chloride Dihydrate
Melting Point
175°C
CAS
10035-04-8
ChemAlert Storage Symbol
Gray
Color
Beige-White
View more