PYREX™ PTFE Product Standard Straight Bore Stopcock Plug Assembly
PYREX™ PTFE Product Standard Straight Bore Stopcock Plug Assembly
Actual product may vary
PYREX™ PTFE Product Standard Straight Bore Stopcock Plug Assembly
Replacement PTFE stopcock plug assemblies for straight bore PTFE stopcocks
Have Questions?
Catalog NumberPlug BoreType
FISC7681-44 mmStopcock Plug Assembly, O-ring Size B
Catalog number FISC7681-4
Price (SGD)
432.00
CS
Add to cart
Plug Bore:
4 mm
Type:
Stopcock Plug Assembly, O-ring Size B
Price (SGD)
432.00
CS
Add to cart
These are replacement Polytetrafluorethylene (PTFE) Product Standard stopcock plug assemblies with various diameter bores for Cat. No. 7282 PYREX straight bore PTFE stopcocks. For more information on PTFE stopcocks, please check the Care of Corning PYREX™ PTFE Stopcocks under Technical Literature.
Specifications
TypeStopcock Plug Assembly, O-ring Size B
Plug Bore4 mm
MaterialPTFE
No. per Pack1/Pk.
No. per Case1/Cs.
For Use With (Equipment)No. 7282 Pyrex straight bore PTFE stopcocks
Unit SizeCS