Bottle Support Base
Actual product may vary
Bottle Support Base
Bottle Support Base
Bottle Support Base
Bottle Support Base
Stabilizes lightweight PTFE and polypropylene bottles
Have Questions?
Catalog number FISBTBASE4
Price (SGD)
-
Four-screw base model secures both round and square bottles.
Accommodates:
  • Round bottles from 50 to 135mm dia.
  • Square bottles from 50 to 135mm diagonal length
Specifications
Product TypeSupport Base
For Use With (Equipment)50 to 135mm dia. round or 50 to 135mm diagonal L square bottles of bottletop dispenser
Width (English)7.52 in.
Width (Metric)191 mm
Height (English)1.77 in.
Height (Metric)44.9 mm
Unit SizeEach