BioCell Vessel and Adapter
Actual product may vary
BioCell Vessel and Adapter
BioCell Vessel and Adapter
BioTek
BioCell Vessel and Adapter
For use with: Epoch, Synergy H1, Synergy HTX.
Have Questions?
Catalog NumberProduct TypeFor Use With (Application)
FISBT7272051Cuvette1cm absorbance measurements
Catalog number FISBT7272051
Price (SGD)
-
Product Type:
Cuvette
For Use With (Application):
1cm absorbance measurements
BioCell Adapter Plate Assembly can hold up to 8 BioCell cuvets (BT7272051) for 1 cm pathlength measurements in a Agilent BioTek microplate spectrophotometer.
Specifications
Product TypeCuvette
For Use With (Equipment)Epoch, Synergy H1, Synergy HTX
For Use With (Application)1cm absorbance measurements
MaterialQuartz
Unit SizeEach