Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Specialty Lab Equipment, Instruments, and Apparatuses
▸
Microplate Instrumentation and Equipment
▸
Microplate Washers and Accessories
▸
Washer Biomagnetic Separation Carrier Accessories
Actual product may vary
BioTek
Washer Biomagnetic Separation Carrier Accessories
Enables high strength biomagnetic separation
Have Questions?
Catalog Number
Product Type
FISBT7102216
96-Well Ring Magnet
Catalog number
FISBT7102216
Price (SGD)
-
Product Type:
96-Well Ring Magnet
Magnet ring
External
For insertion into washer biomagnetic separation carriers
Beads pulled to 4-zone ring at well bottom
Specifications
For Use With (Equipment)
405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Multiflo FX Microplate Washer
Product Type
96-Well Ring Magnet