Phosphate Buffered Saline (PBS), 10X Powder provides a convenient way to make 1 L of 10X stock PBS solution.
Have Questions?
Catalog Number
Quantity
FISBP665-1
2 x 1 L
Catalog number FISBP665-1
Price (SGD)
-
Quantity:
2 x 1 L
Provides a convenient way to make 1 L of 10X stock PBS solution
Eliminates the need to weigh and mix individual components
When reconstituted in ultrapure water such as BP2819, each packet yields 1 L of a 10X PBS solution containing 1.37M NaCl, 0.027M KCl, and 0.119M phosphates
Tested for the absence of DNase, RNase, and protease
PBS is a standard buffer used in many biochemical applications to maintain pH