Water, (For RNA Work) (DEPC-Treated, DNASE, RNASE free/Mol. Biol.), Fisher BioReagents™
Water, (For RNA Work) (DEPC-Treated, DNASE, RNASE free/Mol. Biol.), Fisher BioReagents™
Actual product may vary
Water, (For RNA Work) (DEPC-Treated, DNASE, RNASE free/Mol. Biol.), Fisher BioReagents™
Water, (For RNA Work) (DEPC-Treated, DNASE, RNASE free/Mol. Biol.), Fisher BioReagents™
Water, (For RNA Work) (DEPC-Treated, DNASE, RNASE free/Mol. Biol.), Fisher BioReagents™
DEPC-treated and autoclaved | CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FISBP561-11 LNalgene Poly Bottle
Catalog number FISBP561-1
Price (SGD)
-
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
DEPC-treated and autoclaved
Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work.
Specifications
Chemical Name or MaterialWater
Name NoteFor RNA Work, DEPC-treated and DNASE, RNASE free.
pH7
CAS7732-18-5
ChemAlert Storage SymbolGray
ColorColorless
Melting Point0°C
Boiling Point100°C
Health Hazard 2Low hazard for usual industrial or commercial handling.
Health Hazard 3Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .
NFPA
Health:0
Flammability:0
Instability:0
DNaseDNase free
Molecular FormulaH2O
Merck Index15, 10237
Synonymdihydrogen oxide, dihydrogen monoxide
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
IUPAC Nameoxidane
Molecular Weight (g/mol)18.015
PubChem CID962
ChEBICHEBI:15377
Formula Weight18.02
GradeMolecular Biology
PackagingNalgene Poly Bottle
Vapor Pressure17.5mmHg at 20°C
Filtered ThroughFiltered through a 0.2-micron filter.
Treatment(s)DEPC-Treated
Density1.000g/cm³
Quantity1 L
Physical FormLiquid
Unit SizeEA