Check Order Status
Potassium Phosphate Monobasic (White Crystals), Fisher BioReagents
Element Search
Structure Search
Potassium Phosphate Monobasic (White Crystals), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FISBP362-500
500 g
Glass Bottle
Catalog number
FISBP362-500
Price (SGD)
-
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Phosphate Monobasic
Melting Point
253°C
CAS
7778-77-0
ChemAlert Storage Symbol
Gray
Color
White
View more
This potassium salt is used in buffers for molecular biology and cell culture applications.