Water, Molecular Biology Grade, Fisher BioReagents
Water, Molecular Biology Grade, Fisher BioReagents
Water, Molecular Biology Grade, Fisher BioReagents
Water, Molecular Biology Grade, Fisher BioReagents
Water, Molecular Biology Grade, Fisher BioReagents
For many fundamental molecular biology procedures
Have Questions?
Catalog NumberQuantityPackaging
FISBP2819100100 mLPoly Bottle
Catalog number FISBP2819100
Price (SGD)
-
Quantity:
100 mL
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialWater
Name Note0.03μm filtered to ensure high purity
pH7
CAS7732-18-5
ChemAlert Storage SymbolGray
View more
Ultrapure, deionized, molecular biology grade water is ideal for many fundamental procedures such as PCR, electrophoresis, DNA sequencing, and buffers for enzymatic analyses.