Check Order Status
Rifampicin, Fisher BioReagents™
Element Search
Structure Search
Rifampicin, Fisher BioReagents™
Have Questions?
Catalog Number
Quantity
Packaging
FISBP26791
1 g
Amber Glass
Catalog number
FISBP26791
Price (SGD)
-
Quantity:
1 g
Packaging:
Amber Glass
Request bulk or custom format
Specifications
Chemical Name or Material
Rifampicin
Type
Rifampicin
Physical Form
Solid
CAS
13292-46-1
ChemAlert Storage Symbol
Gray
View more
Rifampicin
Rifampicin is a rifamycin which specifically inhibits DNA-dependent bacterial RNA Polymerase. Mammalian RNA polymerase is not affected.