Check Order Status
Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents
Element Search
Structure Search
Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FISBP2482100
100 mL
Poly Bottle
Catalog number
FISBP2482100
Price (SGD)
-
Quantity:
100 mL
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Ethylenediamine Tetraacetic Acid
Color
Colorless
Name Note
0.5 M Solution, pH 8.0 (for DNA Work)
CAS
60-00-4
ChemAlert Storage Symbol
Gray
View more
EDTA is used extensively in molecular biology to minimize metal ion impurities in reaction buffers. Ideal for DNA work.